Learn More
Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
£27.05 - £63.61
Specifications
Description | RTK Wash Bottle |
---|---|
Material | LDPE |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
10663952
|
Thermo Scientific™
2436-0504 |
500 mL | Isopropanol | ||||||
10014350
|
Thermo Scientific™
2436-0505 |
500 mL | Distilled Water | ||||||
10030681
|
Thermo Scientific™
2436-0506 |
500 mL | Sodium Hypochlorite | ||||||
10024350
|
Thermo Scientific™
2436-0501 |
500 mL | Acetone | ||||||
10070571
|
Thermo Scientific™
2436-1005 |
1000 mL | Distilled Water | ||||||
Description
Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.
- Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
- Unique 38mm colored vented closure designed to prevent dripping and self-dispensing
- Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles) with integrated vent, color coded for easy identification
- Color-coded bar corresponds to closure, preventing closure mix-ups and cross-contamination
- Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
- Flexible LDPE bottle provides soft, ergonomic squeeze
- Sputter-free, precision stream
- Tip can be cut back to provide increased flow
Specifications
RTK Wash Bottle | |
Yes | |
Yes | |
No | |
Cylindrical | |
No |
LDPE | |
Meet OSHA requirements for workplace chemical container labeling. | |
Not Autoclavable | |
Narrow | |
Wash Bottle |