Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 555, Chromotek
Alpaca Recombinant Monoclonal Antibody
Brand:  Chromotek shuGCL555-2-10
 View more versions of this product
Product Code. 18034245
  • £76.00 / 10µL
Explore more special offers
Description
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Specifications
Nano-Secondary™
Alpaca
Secondary
Liquid
CoraLite Plus 555
10 μL
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 555, Chromotek > Quantity: 10 μL