missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets

Unmodified HPTLC silica layers on aluminum for TLC. Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets feature nano-silica 60 on state-of-the-art aluminum sheets that demonstrate low weight, high strength and temperature stability, and easy cutting.

Brand:  Macherey-Nagel™ 818143

211.97 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10185041

This item is not returnable. View return policy

Description

Description

Unmodified HPTLC silica layers on aluminum for TLC. Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets feature nano-silica 60 on state-of-the-art aluminum sheets that demonstrate low weight, high strength and temperature stability, and easy cutting.

Unmodified HPTLC silica layers with High Definition properties.
  • Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents
  • Narrow fractionation of the silica particles allows sharper separations, shorter developing times, shorter migration distances, smaller samples and an increased detection sensitivity compared to SIL G plates
  • Fluorescent Indicator available: manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
Specifications

Specifications

Alugram™ Aluminum
2 to 10 μm
25/Pk.
UV254
0.75 mL/g
∼500 m2/g
20 x 20 cm
Unmodified Silica Gel (SiOH)
60 Å
TLC Plate
HPTLC
Unmodified Silica Gel (SiOH)
0.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers