missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates
Separate mixtures with sharp separations, short developing times and short migration distances. Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates have an increased detection sensitifvity compared to SIL G plates.
£159.00 - £372.00
Specifications
Material | Silica 60 |
---|---|
Particle Size | 2 to 10 μm |
Pore Size | 60 Å |
Type | Glass Plate |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Dimensions (L x W) | Thickness (Metric) | Fluorescent Indicator | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Thickness (Metric) | Fluorescent Indicator | Price | Quantity & Availability | |||
10154991
|
Macherey-Nagel™
811011 |
5 x 5 cm | 0.20 mm | None |
£208.00
Pack of 100 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||
10746322
|
Macherey-Nagel™
811012 |
10 x 10 cm | 0.20 mm | None |
£159.00
Pack of 25 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
10368451
|
Macherey-Nagel™
811021 |
5 x 5 cm | 0.20 mm | UV254 |
£208.00
Pack of 100 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||
10328501
|
Macherey-Nagel™
811022 |
10 x 10 cm | 0.20 mm | UV254 |
£194.00
Pack of 25 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
12780053
|
Macherey-Nagel™
811022 |
10 x 10 cm | 0.20 mm | UV254 | |||||
11814031
|
Macherey-Nagel™
811023 |
10 x 20 cm | 0.20 mm | UV254 |
£372.00
Pack of 50 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
11804031
|
Macherey-Nagel™
811013 |
10 x 20 cm | 0.20 mm | None |
£372.00
Pack of 50 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||
Description
- Nano silica 60, specific surface (BET) ∼ 500 m2/g, mean pore size 60 Å, specific pore volume 0.75 mL/g, particle size 2–10 μm
- Indicator: manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents
Specifications
Silica 60 | |
60 Å | |
0.75 mL/g | |
0.20 mm |
2 to 10 μm | |
Glass Plate | |
∼500 m2/g |