Learn More
Macherey-Nagel™ NANOCOLOR™ COD Tube Tests
Easy, fast COD measurements that conform to ISO 15705 standards. Macherey-Nagel™ Nanocolor™ COD Tube Tests include exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high safety and maximum precision. Available in a range of measurement ranges.
Brand: Macherey-Nagel™ 985023
Additional Details : CAS Number : 7664-93-9 Weight : 0.40000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
Nanocolor™ tube tests feature easy handling for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information at a glance. A range of standardized tube tests are available for ISO 15705-conforming COD analysis using tube tests that are suitable for photometric evaluation. Each tube test is bar coded for automatic recognition by Macherey-Nagel photometers.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
ISO 6060 | |
Determination of chemical oxygen demand (COD) | |
20 x 16mm Cuvettes, pre-filled with reagent and one null cuvette | |
1 Each | |
Potassium dichromate |
Cuvettes | |
Macherey-Nagel Photometers | |
20 | |
1.0 to 10g/L O2 | |
NANOCOLOR Tube Test COD |