Learn More
Macherey-Nagel™ NANOCOLOR™ COD Tube Tests
Easy, fast COD measurements that conform to ISO 15705 standards. Macherey-Nagel™ Nanocolor™ COD Tube Tests include exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high safety and maximum precision. Available in a range of measurement ranges.
£73.00 - £85.00
Specifications
Certifications/Compliance | ISO 6060 |
---|---|
Format | Cuvettes |
For Use With (Application) | Determination of chemical oxygen demand (COD) |
For Use With (Equipment) | Macherey-Nagel Photometers |
Includes | 20 x 16mm Cuvettes, pre-filled with reagent and one null cuvette |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10512523
|
Macherey-Nagel™
985023 |
1.0 to 10g/L O2 |
£85.00
1 test |
Estimated Shipment: 13-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10699452
|
Macherey-Nagel™
985029 |
100 to 1500mg/L O2 |
£73.00
20 tests |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nanocolor™ tube tests feature easy handling for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information at a glance. A range of standardized tube tests are available for ISO 15705-conforming COD analysis using tube tests that are suitable for photometric evaluation. Each tube test is bar coded for automatic recognition by Macherey-Nagel photometers.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
ISO 6060 | |
Determination of chemical oxygen demand (COD) | |
20 x 16mm Cuvettes, pre-filled with reagent and one null cuvette | |
1 Each | |
NANOCOLOR Tube Test COD |
Cuvettes | |
Macherey-Nagel Photometers | |
20 | |
Potassium dichromate |