Learn More
Macherey-Nagel™ NANOCOLOR™ COD Tube Tests
Easy, fast COD measurements that conform to ISO 15705 standards. Macherey-Nagel™ Nanocolor™ COD Tube Tests include exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high safety and maximum precision. Available in a range of measurement ranges.
Brand: Macherey-Nagel™ 985029
Additional Details : CAS Number : 10294-26-5 Weight : 0.07200kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
Nanocolor™ tube tests feature easy handling for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information at a glance. A range of standardized tube tests are available for ISO 15705-conforming COD analysis using tube tests that are suitable for photometric evaluation. Each tube test is bar coded for automatic recognition by Macherey-Nagel photometers.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
ISO 6060 | |
Determination of chemical oxygen demand (COD) | |
20 x 16mm Cuvettes, pre-filled with reagent and one null cuvette | |
1 Each | |
Potassium dichromate |
Cuvettes | |
Macherey-Nagel Photometers | |
20 | |
100 to 1500mg/L O2 | |
NANOCOLOR Tube Test COD |
Safety and Handling
- NANOCOLOR COD 1500
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : sulfuric acid, potassium dichromate, mercury(II) sulfate