missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Nanosep™ Centrifugal Devices with wwPTFE membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
Brand: Cytiva ODPTFE04C34
332.07 GBP valid until 2025-12-31
Use promo code "24320" to get your promotional price.
This item is not returnable.
View return policy
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Clear | |
PTFE (Hydrophilic) | |
Non-sterile | |
100/Pk. |
EFA: 0.3 cm2 | |
0.45 μm | |
Polypropylene | |
50 to 500 μL (Sample) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Nanosep™ Centrifugal Devices with wwPTFE membrane > 0.45 μm; 100/Pk.
Spot an opportunity for improvement?Share a Content Correction