missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Nanosep™ Centrifugal Devices with wwPTFE membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
£358.00 - £1600.00
Specifications
Color | Clear |
---|---|
Description | EFA: 0.3 cm2 |
Material (Membrane) | wwPTFE |
Sterility | Non-sterile |
Housing Material | Polypropylene |
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
17174801
|
Cytiva (formerly Pall Lab)
ODPTFE02C34 |
0.2 μm | 100/Pk. |
£671.83
Pack of 100 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17184801
|
Cytiva (formerly Pall Lab)
ODPTFE02C35 |
0.2 μm | 500/Pk. |
£1600.00
Pack of 500 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
17194801
|
Cytiva (formerly Pall Lab)
ODPTFE04C34 |
0.45 μm | 100/Pk. |
£358.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
17104811
|
Cytiva (formerly Pall Lab)
ODPTFE04C35 |
0.45 μm | 500/Pk. |
£1430.00
Pack of 500 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Clear | |
wwPTFE | |
Polypropylene |
EFA: 0.3 cm2 | |
Non-sterile | |
50 to 500 μL (Sample) |