Learn More
Emma Safety Footwear Napoli Safety Shoes
Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks allow you to move easily between the office and warehouse.
Brand: Emma Safety Footwear MM11109044
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Nitrile Rubber Heat Resistant Outsole
- Full grain leather/Nubuck
- ESD Certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Brown | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
44 |
Laces | |
Napoli | |
Leather, Nubuck | |
Steel |