Learn More
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC analyzers
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
Brand:  Thermo Scientific™ C4010-1296
 View more versions of this product
Product Code. 12403628
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
  • Maximum fill volume: 5mL
  • Superior quality N-51A clear borosilicate glass
  • 5mL sample vial is designed for use in the Shimadzu™ Model ASI-5000
  • Screw thread vial fits standard 10-425 closures (included with pre-assembled product)
  • Open-top vial was designed in cooperation with the original equipment manufacturer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Thread TOC Vial, Polypropylene Cap with PTFE septum
5 mL
10-425
Shimadzu Model ASI-5000