Learn More
Thermo Scientific™ Assembled Target™ 10-425 Caps and Septa
Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.
Brand:  Thermo Scientific™ C4010-60AY
 View more versions of this product
Product Code. 12483658
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yellow; Septa: Red PTFE/White Silicone
PTFE, Silicone, Polypropylene
National Assembled Target
10 mm
Yellow
Red PTFE/White Silicone
Screw Cap
10-425 Screw Thread Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.