missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.

Brand:  Thermo Scientific™

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12478666

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.

  • Unassembled convenience kits
  • Choice of 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass
  • Eliminate leaching of ions
  • Provide consistent pH for duration of sample storage life
  • With polypropylene open top screw caps and PTFE/silicone septa
  • Convenient packaging for storerooms or laboratory cabinets
Specifications

Specifications

Amber
Glass
PTFE/Silicone
Screw Cap (Open-top) with Septum
18-400
21 mm
16 mL
4.375 dr.
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers