missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Standard-Opening 12 x 32mm Screw-Thread Glass Vials

Thermo Scientific™ National Standard-Opening Screw-Thread Glass Vials feature a uniformly flat bottom for security with inserts.

Brand:  Thermo Scientific™ C4013-K1W

 View more versions of this product

Product Code. 12460158

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ National Standard-Opening Screw-Thread Glass Vials feature a uniformly flat bottom for security with inserts.

  • Standard neck vials with 8-425 thread finish
  • Superior quality borosilicate clear glass Type 1, Class A or 51A amber glass
  • I-D™ vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Deactivated vials are Silanized or Kimshield™ pretreated
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2mL Clear I-D, Kimshield
Glass
1.8 mL
Yes
Clear
2 mL
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers