Learn More
Thermo Scientific™ National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
Brand:  Thermo Scientific™ C4000-80W
 View more versions of this product
Product Code. 10558415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
  • Convenience Kits with 2mL, 12 × 32mm vials
  • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Blue caps with your choice of vial and preassembled seals with septa
  • Packaged in two-compartment benchtop storage tray
  • All items remain dust-free
  • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
Clear
2 mL
9-425
Clear Glass with I-D; PTFE/Red Rubber septa
PTFE, Rubber
PTFE/red rubber
Yes