Learn More
Thermo Scientific™ National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
£27.17 - £50.20
Specifications
Closure Color Blue
Closure Size 9-425
View More Specs
Products 4
Product Code Brand dimType Closure Color Septum Color Price Quantity & Availability  
Product Code Brand dimType Closure Color Septum Color Price Quantity & Availability  
12403538
View Documents Promotion Details
Thermo Scientific™
C4000-80
Blue PTFE/red rubber Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10558415
View Documents Promotion Details
Thermo Scientific™
C4000-80W
Blue PTFE/red rubber Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11978825
View Documents Promotion Details
Thermo Scientific™
C400092
Blue Red PTFE/White Silicone Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11971563
View Documents Promotion Details
Thermo Scientific™
C4000-97
Blue Red PTFE/White Silicone Natural This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
  • Convenience Kits with 2mL, 12 × 32mm vials
  • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Blue caps with your choice of vial and preassembled seals with septa
  • Packaged in two-compartment benchtop storage tray
  • All items remain dust-free
  • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
Specifications
Specifications
Blue
9-425