Learn More
Deutsch Neumann™ Rubber Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ Rubber Safety Pipette Fillers are manufactured from natural rubber.
Brand:  Deutsch Neumann™ 4700001
 View more versions of this product
Product Code. 10313441
  • £4.90 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Only two operating points
  • Excellent tensile strength and resilience
  • One-handed design is the simplest safety pipette available
Specifications
Specifications
Red
Standard 1
For comfortable pipetting toxic, corrosive, containing bacteria and malodorous substances
Natural Rubber
Safety Pipette Filler
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Deutsch Neumann™ Rubber Safety Pipette Fillers > Standard 1 Model