Learn More
Abeba™ Nature 4040 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 4040/42
71.23 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : CAS Number : 161233-97-2 Weight : 1.20000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
- Resistant to oil and benzine
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
42 | |
Rubber |
White | |
Leather | |
Slip-resistant | |
4040 |