Learn More
Abeba™ Nature 4055 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 4055/45
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
- Resistant to oil and petrol
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
45 | |
Rubber |
Black | |
Leather | |
Slip-resistant | |
4055 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Abeba™ Nature 4055 Shoes > Size: 45
Spot an opportunity for improvement?Share a Content Correction