Learn More
Abeba™ Nature 8040 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 8040/47
Additional Details : Weight : 0.80000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
47 | |
Rubber |
White | |
Leather | |
Slip-resistant | |
8040 |