Learn More
Abeba™ Nature 8040 Shoes
Leather covered cork footbed is ESD dissipative.
£72.81 - £79.87
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC |
---|---|
Color | White |
Footbed Material | Cork |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15682968
|
Abeba™
8040/34 |
34 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15692968
|
Abeba™
8040/35 |
35 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15602978
|
Abeba™
8040/36 |
36 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15612978
|
Abeba™
8040/37 |
37 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15622978
|
Abeba™
8040/38 |
38 |
£72.81
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15632978
|
Abeba™
8040/39 |
39 |
£72.81
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15642978
|
Abeba™
8040/40 |
40 |
£79.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15652978
|
Abeba™
8040/41 |
41 |
£72.81
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15662978
|
Abeba™
8040/42 |
42 |
£72.81
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15672978
|
Abeba™
8040/43 |
43 |
£72.81
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15682978
|
Abeba™
8040/44 |
44 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15692978
|
Abeba™
8040/45 |
45 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15602988
|
Abeba™
8040/46 |
46 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15612988
|
Abeba™
8040/47 |
47 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15622988
|
Abeba™
8040/48 |
48 |
£72.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
Slip-resistant | |
8040 |
White | |
Leather | |
Rubber |