Learn More
Abeba™ Nature 8040 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 8040/35
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15692968
  • £72.81 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
35
Rubber
White
Leather
Slip-resistant
8040