missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NAV 2729
Selective ARF6 inhibitor
Brand: Tocris Bioscience™ 5986/10
158.96 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
NAV 2729 is a selective ARF6 inhibitor (IC50 = 1.0 μM). Selective for ARF6 over other ARF family members and small GTPases. Inhibits ARF6 activation in uveal melanoma cells in vitro and blocks downstream signaling pathways of oncogenic GNAQ. Reduces tumorigenesis in mouse model of uveal melanoma.Specifications
Yellow | |
419547-11-8 | |
456.88g/mol | |
Store at 4°C | |
10 mg |
Inhibitor | |
C25H17ClN4O3 | |
Soluble to 100mM in DMSO | |
Blocks downstream signaling pathways |
For Research Use Only