missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NE-PER™ Nuclear and Cytoplasmic Extraction Reagents

Efficiently lyse cells and extract separate cytoplasmic and nuclear protein fractions in less than two hours with this subcellular fractionation kit.

Brand:  Thermo Scientific™ 78835

426.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10309383

  • £600.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ NE-PER Nuclear and Cytoplasmic Extraction Kit provides for efficient cell lysis and extraction of separate cytoplasmic and nuclear protein fractions in less than two hours.

The NE-PER Kit is a nuclear protein extraction method that involves simple, stepwise lysis of cells and centrifugal isolation of nuclear and cytoplasmic protein fractions. A benchtop microcentrifuge, tubes and pipettors are the only tools required. The NE-PER Reagents efficiently solubilize and separate cytoplasmic and nuclear proteins into fractions with minimal cross-contamination or interference from genomic DNA and mRNA. Once desalted or diluted, the isolated proteins can be used to perform immunoassays and protein interaction experiments, such as mobility shift assays (EMSA), co-immunoprecipitation (Co-IP) and pull-down assays.

Highlights:

  • Fast – obtain nuclear and cytoplasmic fractions in less than two hours
  • Proven – the NE-PER Kit is referenced in more than 950 distinct publications
  • Scalable – two kit sizes for producing extracts from cells and tissues
  • Convenient – simple instructions do not require ultracentrifugation over gradients
  • Compatible – use for downstream assays, including Western blotting, gel-shift assays, protein assays, reporter gene assays and enzyme activity assays

Includes:

Two kit sizes, each containing three buffer and detergent solutions

Compatible with:

Thermo Scientific LightShift Chemiluminescent EMSA Kit (Mfr. No. 20148)

Storage Requirements:

Upon receipt store kit components at 4°C

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nuclear and Cytoplasmic Extraction Reagent
Ambient
NE-PER™
Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation
Upon receipt store kit components at 4°C. Product is shipped at ambient temperature.
Liquid
Lysis Buffer
75 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NE-PER Nuclear and Cytoplasmic Extraction Reagents

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : non ionic detergent

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.