Learn More
Thermo Scientific™ NE-PER™ Nuclear and Cytoplasmic Extraction Reagents
Efficiently lyse cells and extract separate cytoplasmic and nuclear protein fractions in less than two hours with this subcellular fractionation kit.
Brand: Thermo Scientific™ 78833
274.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : CAS Number : 813-19-4 Weight : 0.80000kg
Description
Thermo Scientific™ NE-PER Nuclear and Cytoplasmic Extraction Kit provides for efficient cell lysis and extraction of separate cytoplasmic and nuclear protein fractions in less than two hours.
The NE-PER Kit is a nuclear protein extraction method that involves simple, stepwise lysis of cells and centrifugal isolation of nuclear and cytoplasmic protein fractions. A benchtop microcentrifuge, tubes and pipettors are the only tools required. The NE-PER Reagents efficiently solubilize and separate cytoplasmic and nuclear proteins into fractions with minimal cross-contamination or interference from genomic DNA and mRNA. Once desalted or diluted, the isolated proteins can be used to perform immunoassays and protein interaction experiments, such as mobility shift assays (EMSA), co-immunoprecipitation (Co-IP) and pull-down assays.
Highlights:
- Fast – obtain nuclear and cytoplasmic fractions in less than two hours
- Proven – the NE-PER Kit is referenced in more than 950 distinct publications
- Scalable – two kit sizes for producing extracts from cells and tissues
- Convenient – simple instructions do not require ultracentrifugation over gradients
- Compatible – use for downstream assays, including Western blotting, gel-shift assays, protein assays, reporter gene assays and enzyme activity assays
Includes:
Two kit sizes, each containing three buffer and detergent solutions
Compatible with:
Thermo Scientific LightShift Chemiluminescent EMSA Kit (Mfr. No. 20148)
Storage Requirements:
Upon receipt store kit components at 4°C
Specifications
Nuclear and Cytoplasmic Extraction Reagent | |
Ambient | |
NE-PER™ | |
Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation |
Upon receipt store kit components at 4°C. | |
Liquid | |
Lysis Buffer | |
15 mL |
Safety and Handling
- NE-PER Nuclear and Cytoplasmic Extraction Reagents
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Non-ionic Detergent,Salt
For Research Use Only. Not for use in diagnostic procedures.