missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17906-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Involved in endocytosis.Specifications
NECAP1 | |
Polyclonal | |
Unconjugated | |
NECAP1 | |
NECAP 1, NECAP endocytosis associated 1, NECAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25977, 312694, 67602 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P69682, Q8NC96, Q9CR95 | |
Necap1 | |
NECAP1 Fusion Protein Ag12283 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |