missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | NECAP1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16817914
|
Proteintech
17906-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16807914
|
Proteintech
17906-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Involved in endocytosis.Specifications
NECAP1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P69682, Q8NC96, Q9CR95 | |
25977, 312694, 67602 | |
NECAP1 Fusion Protein Ag12283 | |
Primary | |
-20°C | |
NECAP1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NECAP 1, NECAP endocytosis associated 1, NECAP1 | |
Necap1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |