Learn More
Dr.Weigert™ Neodisher FA
Neodisher LaboClean FA - Alkaline detergent, liquid concentrate for the automated cleaning of surgical thermo-stable and thermolabile instruments and others medicals or laboratories utensils. Versatile and concentrated detergent.
Brand: Dr.Weigert™ 410135
Additional Details : CAS Number : 6834-92-0 Weight : 7.40000kg Transport : UN number : 1719 Chem class : 8 Pack group : III
Description
- Alkaline detergent, liquid concentrate
- neodisher LaboClean FA is used for cleaning in washer disinfection
- Free of surfactants and active chlorine Excellent material compatibility
- Cleans blood, protein, culture media and gelatine
- Dosage between 2-4 mL/l
Specifications
Neodisher FA | |
11.6 to 11.8 | |
Disodium metasilicate, potassium silicate, potassium hydoxide | |
Cannister | |
250 x 190 x 140 mm | |
5 L |
Liquid | |
Non-sterile | |
1 to 4 mL/L | |
1.5 g/cm3 (20°C) | |
Filtered |