Learn More
Dr.Weigert™ Neodisher N
Neodisher N - Acidic detergent and neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent. Automated acidic cleaning of animal cages.
Brand: Dr.Weigert™ 420187
Additional Details : Weight : 1.47000kg Transport : UN number : 1805 Chem class : 8 Pack group : III
Description
- Based on phosphoric acid
- neodisher N is used for neutralizing alkaline residues in washer disinfection
- Available in either bottles or canisters
- Has very good rinsing properties
- Suitable for use with: surgical instruments, anesthesia items, containers, surgical footwear, baby bottles, and other medical devices, as well as laboratory glassware Free of surfactantsDosage of between 2-4 mL/L
- Titration factor: 0.16
Specifications
Neodisher N | |
2.2 to 1.8 | |
Phosphoric acid, citric acid | |
Bottle | |
Neutralizing Any Carried Over Alkaline Residues on Laboratory Glassware or for Acid Pre-Cleaning Resp | |
12/Pk. |
Liquid | |
Non-sterile | |
1 to 4 mL/L | |
250 x 100 x 60 mm | |
12/Pk. | |
1 L |
Safety and Handling
- neodisher N
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.