Learn More
Dr.Weigert™ Neodisher N
Neodisher N - Acidic detergent and neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent. Automated acidic cleaning of animal cages.
Brand: Dr.Weigert™ 420133
Additional Details : CAS Number : 7664-38-2 Weight : 7.35000kg Transport : UN number : 1805 Chem class : 8 Pack group : III
Description
- Based on phosphoric acid
- neodisher N is used for neutralizing alkaline residues in washer disinfection
- Available in either bottles or canisters
- Has very good rinsing properties
- Suitable for use with: surgical instruments, anesthesia items, containers, surgical footwear, baby bottles, and other medical devices, as well as laboratory glassware Free of surfactantsDosage of between 2-4 mL/L
- Titration factor: 0.16
Specifications
Neodisher N | |
2.2 to 1.8 | |
Phosphoric acid, citric acid | |
Canister | |
Filtered | |
5 L |
Liquid | |
Non-sterile | |
1 to 4 mL/L | |
220 x 240 x 140 mm | |
Neutralizing Any Carried Over Alkaline Residues on Laboratory Glassware or for Acid Pre-Cleaning Resp |