missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher Z

Neodisher Z - Acidic neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent during automated reprocessing. Free of surfactants and phosphates.

Brand:  Dr.Weigert™ 420287

Additional Details : Weight : 1.22000kg


 View more versions of this product

Product Code. 10308807

  • £176.00 / Pack of 12
Estimated Shipment: 21-08-2024
to see stock.

Description

Description

  • Based on citric acid
  • neodisher Z is used for neutralizing alkaline residues in washer disinfection
  • Without phosphates, phosphoric acid, nitrogens compounds and surfactants
  • Dosage of between 2-4 mL/L
  • Gentle on materials
  • Laboratory glassware in medical, industrial and water laboratories
Specifications

Specifications

Neodisher Z
3 to 2.6
Citric acid, 2-octyl-2 H-isothiazol-3-one
1.2 g/cm3 (20°C)
Filtered
Liquid
Non-sterile
1 to 5 mL/L
250 x 100 x 60 mm
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • neodisher Z

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : citric acid, anhydrous

SDS
Documents

Documents

Certificates
Special Offers

Special Offers