Learn More
Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Blue |
Physical Form | Wet Crystalline Aggregates |
Quantity | 100 g |
Assay Percent Range | 99.99% (REO) |
IUPAC Name | neodymium(3+);trinitrate;hydrate |
Formula Weight | 330.25 (Anhydrous) |
Odor | Odorless |
Appearance | Violet crystals |
Chemical Name or Material | Neodymium(III) nitrate hydrate |
Product Suggestions
Customers who viewed this item also viewed
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.