missing translation for 'onlineSavingsMsg'
Learn More
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals Product Code.: 15471967

Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals

Product Code. 15471967
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15471967

Brand: Thermo Scientific Alfa Aesar 044181.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray to Silver
UN Number UN3089
Quantity 5 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Neodymium powder
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT14I004 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number044181.14,044182.14
Lot NumberU10H028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044181.14,044182.14
Lot NumberC16R031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044181.14,044182.14
Lot NumberD25Q35 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044181.14,044182.14
Lot NumberQ25F017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044181.14,044182.14
compliance-icons compliance-icons
Product Identifier
  • Neodymium powder, -40 mesh, 99.8% (REO)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H251-Self-heating: may catch fire.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.
  • P240-Ground/bond container and receiving equipment.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P407-Maintain air gap between stacks/pallets.
  • P420-Store away from other materials.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Neodymium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.