missing translation for 'onlineSavingsMsg'
Learn More
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals Product Code.: 15481967

Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals

Product Code. 15481967
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15481967

Brand: Thermo Scientific Alfa Aesar 044181.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray to Silver
UN Number UN3089
Quantity 25 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Neodymium powder
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT14I004 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number044181.14,044182.14
Lot NumberU10H028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044181.14,044182.14
Lot NumberC16R031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044181.14,044182.14
Lot NumberD25Q35 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044181.14,044182.14
Lot NumberQ25F017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044181.14,044182.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.