missing translation for 'onlineSavingsMsg'
Learn More
Neodymium ingot, 99.1% (metals basis), Thermo Scientific Chemicals Product Code.: 11310699

Neodymium ingot, 99.1% (metals basis), Thermo Scientific Chemicals

Product Code. 11310699
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11310699

Brand: Thermo Scientific Alfa Aesar 000214.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray-Silver
Quantity 100 g
Percent Purity 99.1%
Form Ingot
Assay (metals basis)
Chemical Name or Material Neodymium ingot

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW16F009 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog Number000214.14,000214.22
Lot NumberU06K042 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number000214.14,000214.22
Lot NumberS18K002 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number000214.14,000214.22
Lot NumberQ04K001 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number000214.14,000214.22
Lot NumberX25J042 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number000214.14,000214.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium ingot, 99.1% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.