Learn More
Invitrogen™ Neon™ NxT Electroporation System 10-μL Kit with 1-Channel Tubes
The Neon NxT Electroporation System 10-μL Kit is designed specifically for use with the Neon NxT Electroporation System.
Brand: Invitrogen™ N1096
1789.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
The Neon NxT Electroporation System 10-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 10 μL, containing 5 x 104–2 x 105 adherent cells or 1 x 105–5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be washed and plated into a multi-well culture dish.
Specifications
• 4 x 1 mL Resuspension R Buffer; store at 4°C • 4 x 1 mL Resuspension T Buffer; store at 4°C • 4 x 1 mL Resuspension Genome Editing Buffer; store at 4°C • 2 x 100 mL Electrolytic E10 Buffer; store at 4°C • 96 reaction tips • 32 electroporation 1-channel tubes |
|
Electroporation Pipette Tip | |
Neon™ NxT Electroporation System | |
96 x 2 Reactions | |
96 x 2 reactions |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Electroporation | |
Transfection | |
Pipette Tip | |
10 μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.