Learn More
Invitrogen™ Neon™ NxT Electroporation System 100-μL Kit
The Neon NxT Electroporation System 100-μL Kit is designed specifically for use with the Neon NxT Electroporation System.
Brand: Invitrogen™ N10025
621.30 GBP valid until 2024-06-28
Use promo code "21689" to get your promotional price.
Additional Details : Weight : 0.40000kg
Description
The Neon NxT Electroporation System 100-μL Kit is designed specifically for use with the Neon NxT Electroporation System. Use this kit for transfection volumes of 100 μL, containing 5 x 105–2 x 106 adherent cells or 1 x 106–5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be washed and plated into a multi-well culture dish.
Specifications
• 10 mL Resuspension R Buffer; store at 4°C • 10 mL Resuspension T Buffer; store at 4°C • 10 mL Resuspension Genome Editing Buffer; store at 4°C • 50 mL Electrolytic E100 Buffer; store at 4°C • 25 reaction tips • 8 electroporation tubes |
|
Electroporation Pipette Tip | |
Neon™ NxT Electroporation System | |
25 x 2 Reactions | |
100 μL |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Electroporation | |
Transfection | |
Pipette Tip |
For Research Use Only. Not for use in diagnostic procedures.