Learn More
Invitrogen™ Neon™ NxT Electroporation System 100-μL Kit with 1-Channel Tubes
The Neon NxT Electroporation System 100-μL Kit is designed specifically for use with the Neon NxT Electroporation System.
Brand:  Invitrogen™ N10096
 View more versions of this product
Product Code. 17819444
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Neon NxT Electroporation System 100-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 100 μL, containing 5 x 105–2 x 106 adherent cells or 1 x 106–5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be washed and plated into a multi-well culture dish.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 4 x 10 mL Resuspension R Buffer; store at 4°C
• 4 x 10 mL Resuspension T Buffer; store at 4°C
• 4 x 10 mL Resuspension Genome Editing Buffer; store at 4°C
• 2 x 100 mL Electrolytic E100 Buffer; store at 4°C
• 96 reaction tips
• 32 electroporation 1-channel tubes
Electroporation Pipette Tip
Neon™ NxT Electroporation System
96 x 2 Reactions
96 x 2 reactions
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Electroporation
Transfection
Pipette Tip
100 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Neon™ NxT Electroporation System 100-μL Kit with 1-Channel Tubes > 96 x 2 Reactions