missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neon™ Transfection System 100 μL Kit
Neon™ Transfection System 100 μL Kit
Brand: Invitrogen™ MPK10025
542.65 GBP valid until 2024-06-28
Use promo code "21689" to get your promotional price.
Code : 50
Additional Details : Weight : 0.56000kg
Description
The Neon™ Transfection System 100 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 100 μL, containing 5 × 105–2 × 106 adherent cells or 1 × 106–5 × 106 suspension cells. Cells that have been transfected using the included 100μL Neon™ Tips are then ready to be washed and plated into a 6-well, 60 mm, or 10 cm culture dish, depending on your protocol.Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum | |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA | |
Electroporation | |
Transfection | |
Neon™ | |
25 x 2 Reactions | |
Room Temperature | |
Neon™ Transfection System |
The Neon™ Transfection System 100 μL Kit comes with 10 mL resuspension buffer R, 10 mL resuspension buffer T, and 75 mL E2 electrolytic buffer in one box and 25 x 100μL reaction tips and 5 electroporation tubes in a separate box. Refer to the Neon™ Transfection System users manual for protocol instructions. Store the buffer box at 4°C and the tips & tubes box at room temperature. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
CMV | |
Neon™ Transfection System | |
50 Reactions | |
Transfection System | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type | |
100μl |
For Research Use Only. Not for use in diagnostic procedures.