missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neon™ Transfection Tubes
The Neon™ Transfection Tubes are designed specifically for use with the Neon™ Transfection System device (MPK5000).
Brand: Invitrogen™ MPT100
This item is not returnable.
View return policy
Description
Each Neon™ Transfection System Kit (MPK1096 and MPK10096) already comes with 20 electroporation tubes. The additional pack of 100 tubes is offered for those labs wishing to use single tubes per reaction depending on their experimental set-up.
Specifications
The Neon™ Transfection Tubes comes in a pack of 100 tubes. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions. | |
RNA, DNA | |
Transfection | |
Transfection Tube | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Neon™ Transfection System | |
Neon™ | |
1 Pack | |
Room Temperature | |
Neon™ Transfection System |
User Bulletin: CRISPR/Cas9-mediated Genome Editing of Human Primary T-cells using the Neon Transfection System
Version: A.0
File size: 104.9 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Neon™ Transfection Tubes > Transfection Tubes (100)
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction