Learn More
Fisherbrand™ Neoprene Tubing
Used in flow lines for fluids and gases in a variety of applications.
£15.68 - £440.00
Specifications
Material Neoprene
Overall Length (Metric) 10 m
View More Specs
Products 13
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11885492
View Documents
Fisherbrand™
FB50781
10 x 12mm
£45.45
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11577063
View Documents
Fisherbrand™
FB50811
10 x 24mm
£203.00
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11895492
View Documents
Fisherbrand™
FB50783
12.5 x 17mm
£57.55
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11587063
View Documents
Fisherbrand™
FB50813
12.5 x 29.5mm
£440.00
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11825502
View Documents
Fisherbrand™
FB50789
25 x 31.5mm
£161.00
10m
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
11845492
View Documents
Fisherbrand™
FB50773
3 x 4.5mm
£15.68
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11835502
View Documents
Fisherbrand™
FB50803
3 x 9mm
£35.35
10m
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
11845502
View Documents
Fisherbrand™
FB50805
5 x 13mm
£157.02
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11855492
View Documents
Fisherbrand™
FB50775
5 x 8mm
£43.06
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
10366344
View Documents
Fisherbrand™
FB50777
6.5 x 1.5mm
£80.47
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11875492
View Documents
Fisherbrand™
FB50779
8 x 10mm
£125.00
10m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11865502
View Documents
Fisherbrand™
FB50809
8 x 20mm
£343.20
10m
In Stock
to see stock.
Edge
Add to basket
11855502
View Documents
Fisherbrand™
FB50807
6.5 x 10mm
£280.49
10m
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
  • Resistant to heat, ozone, petroleum oils and hydrocarbon solvents
  • Temperature range: -30°C to 100°C
Specifications
Specifications
Neoprene
10 m
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Neoprene Tubing