Learn More
Lovibond Tintometer™ Nessleriser Comparator
Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.
Brand:  Lovibond Tintometer™ 172250
 View more versions of this product
Product Code. 13121671
  • £304.00 / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.
  • Color comparator/colorimeter
  • Usess comparator disc standards
  • Determines chemical concentrations in solutions
  • Available options:
    • Tube length (250mm/113mm)
    • Lighting (lighting unit or ambient light)
    • DB 420 glass anti-meniscus plungers
Specifications
Specifications
Nessleriser 2250
Stand