missing translation for 'onlineSavingsMsg'
Learn More

Lovibond Tintometer™ Nessleriser Comparator

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.


Products 4
Product Code Brand Description Includes Price Quantity & Availability  
Product Code Brand Description Includes Price Quantity & Availability  
13131671
View Documents
Lovibond Tintometer™
172170
Nessleriser 2250 Upgrade Kit Pair Nessler tubes, 250 mm with plungers N/A
N/A
13121671
View Documents
Lovibond Tintometer™
172250
Nessleriser 2250 Stand N/A
N/A
13191661
View accessory products View Documents
Lovibond Tintometer™
172030
Nessleriser 2150 Stand, daylight lighting unit and pair Nessler tubes, 113 mm with plungers N/A
N/A
13101671
View accessory products View Documents
Lovibond Tintometer™
172040
Nessleriser 2250 Stand, daylight lighting unit and pair Nessler tubes, 250 mm with plungers N/A
N/A
Description

Description

Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.

  • Color comparator/colorimeter
  • Usess comparator disc standards
  • Determines chemical concentrations in solutions
  • Available options:
    • Tube length (250mm/113mm)
    • Lighting (lighting unit or ambient light)
    • DB 420 glass anti-meniscus plungers
Videos
SDS
Documents

Documents

Product Certifications