Learn More
Eppendorf™ Scientific shaker Microplate Platform
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand: Eppendorf™ TTR-221
This item is not returnable.
View return policy
Description
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Microplate Rack | |
Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series | |
5 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Eppendorf™ Scientific shaker Microplate Platform > Microplate rack; For single layer of 5 plates
Spot an opportunity for improvement?Share a Content Correction