missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for Digital Microplate Shakers

Get assorted accessories for Digital Microplate Shakers

Brand:  Thermo Scientific™ 88882116

 View more versions of this product

Product Code. 16514802

  • £140.16 / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Assorted accessories for Digital Microplate Shakers

Service Plan

Standard Thermo warranty guideline, provide repair service and free spare part within warranty

Services

2 years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Platform
Stainless Steel
11.49 in.
10.82 in.
Platform Large Assemble
Large
Certificate of Analysis (COA), Certificate of Conformance (COC)
292 x 275 x 25 mm
292 mm
275 mm
Rocker, Rotator, mixer
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Accessories for Digital Microplate Shakers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.