missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NHS Mag Sepharose™

NHS Mag Sepharose magnetic beads for convenient enrichment of selected proteins using pull-down sample preparation techniques

Brand:  Cytiva 28-9440-09

 View more versions of this product

Product Code. 10371315

  • £42.42 / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity. An antibody or other protein with desired affinity is covalently coupled to the medium via primary amines for the purpose of capture of target proteins of interest.

  • Visible and dense Sepharose based magnetic beads with N-hydroxysuccinimide (NHS) ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/pull-down of target proteins - only low amounts of bound capture molecule
  • Flexible protocols with elution conditions optimized for downstream applications such as electrophoresis and MS

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: N-hydroxysuccinimide
Ligand density: 8-14μmol/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% 2-propanol
Storage temperature: 4°C to 8°C

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NHS Mag Sepharose
8 to 14μM/mL
37 to 100 μm
4°C to 8°C
N-hydroxysuccinimide
8 to 14μM/mL Gel
Paramagnetic spherical, highly cross linked agarose particles
500 μL
For pull-down techniques enabling rapid capture and enrichment of selected Proteins based on affinity
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

SDS
Documents

Documents

Certificates
Special Offers

Special Offers