Learn More
Cytiva Ni Sepharose™ High Performance Media
Precharged with Ni2+ for optimal purification of histidine-tagged proteins. Cytiva Lifescience™ Ni Sepharose™ High Performance Media shows high binding capacity and compatibility.
Brand: Cytiva 17-5268-02
Code : GE
Additional Details : Weight : 0.17100kg
Description
- Negligible leakage of Ni2+ ions
- High binding capacity: at least 40mg protein/mL medium
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Stable (long-term) in the pH range 3-12
- Average particle size: 34μm
Specifications
Ni Sepharose HP chromato graphy media | |
3 to 12 | |
6% Cross-linked Agarose | |
Ni-Sepharose |
100mL | |
34μm | |
4° to 30°C |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains Nickel. May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, Nickel