missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel 201 Laboratory Dishes

Nickel Electro™ Nickel 201 Laboratory Dishes are made to resist corrosive enviroments, which make it useful for maintaining product purity.

Brand:  Nickel Electro™ 503

 View more versions of this product

Product Code. 12777356

This item is not returnable. View return policy

Description

Description

  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications

Specifications

Flat
Nickel
Evaporating Dish
16 mm
45 mL
Round
Bright
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers