missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel 201 Laboratory Dishes

Nickel Electro™ Nickel 201 Laboratory Dishes are made to resist corrosive enviroments, which make it useful for maintaining product purity.

Brand:  Nickel Electro™ 504

 View more versions of this product

Product Code. 12787356

  • £33.40 / Each
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications

Specifications

Flat
Nickel
Evaporating Dish
19 mm
73 mL
Round
Bright
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers