missing translation for 'onlineSavingsMsg'
Learn More

Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

£37.19 - £372.86

Chemical Identifiers

CAS 207861-63-0
Molecular Formula C4F12N2NiO8S4
Molecular Weight (g/mol) 618.964
MDL Number MFCD23380174
InChI Key VFUSIQAHVPEYFD-UHFFFAOYSA-N
Synonym nickel bis-trifluoromethanesulfonimide
PubChem CID 131875825
IUPAC Name bis(trifluoromethylsulfonyl)azanide;nickel(2+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15412847
View Documents
Thermo Scientific Alfa Aesar
047102.MD
250 mg
£37.19
250mg
Please to purchase this item. Need a web account? Register with us today!
15492837
View Documents
Thermo Scientific Alfa Aesar
047102.03
1 g
£104.52
1g
Please to purchase this item. Need a web account? Register with us today!
15402847
View Documents
Thermo Scientific Alfa Aesar
047102.06
5 g
£372.86
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207861-63-0
618.964
VFUSIQAHVPEYFD-UHFFFAOYSA-N
131875825
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
C4F12N2NiO8S4
MFCD23380174
nickel bis-trifluoromethanesulfonimide
bis(trifluoromethylsulfonyl)azanide;nickel(2+)

Specifications

207861-63-0
Solid
[(CF3SO2)2N]2Ni
UN3261
It is soluble in water.
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]
618.964
618.96
Nickel bis(trifluoromethylsulfonyl)imide
Green
C4F12N2NiO8S4
MFCD23380174
nickel bis-trifluoromethanesulfonimide
VFUSIQAHVPEYFD-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;nickel(2+)
131875825
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H314-H318-H317
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335-H350

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only